Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547349
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547349 Supplier Invitrogen™ Supplier No. PA547349
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) ILT2, rhILT4, ILT5 is observed, and less than 5% cross-reactivity with rhLIR5 and rhLIR7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades (see LILRA1, MIM 604810). One member of subfamily A (LILRA3) lacks a transmembrane region and is presumed to be a soluble receptor (Borges et al., 1997 [PubMed 9548455]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA3
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LILRA3
Gene Accession No. Q8N6C8
Gene Alias CD85 antigen-like family member E; CD85e; HM31; HM43; ILT6; ILT-6; Immunoglobulin-like transcript 6; leucocyte Ig-like receptor A3; leukocyte immunoglobulin like receptor A3; leukocyte immunoglobulin-like receptor 4; leukocyte immunoglobulin-like receptor A3; leukocyte immunoglobulin-like receptor subfamily A member 3; leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3; LILRA3; LIR4; LIR-4; Monocyte inhibitory receptor HM43/HM31
Gene Symbols LILRA3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA3/CD85e Arg18-Glu439.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11026
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.