Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA5 Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA531314
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531314 Supplier Invitrogen™ Supplier No. MA531314
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat LILRA5.

Leukocyte immunoglobulin-like receptor A5 (LILRA5) is a gene located on chromosome 19q13.42, encoding a protein that is part of the leukocyte immunoglobulin-like receptor (LILR) family. These receptors are primarily expressed on immune cells, such as natural killer cells, monocytes, and dendritic cells, and play a pivotal role in modulating immune responses. LILRA5 is thought to function as an activating receptor, influencing the activation and regulation of immune cell functions. It is implicated in processes such as antigen presentation, cytokine production, and phagocytosis, contributing to both innate and adaptive immune responses. Dysregulation of LILRA5 expression or function can impact immune homeostasis and has been associated with various autoimmune and inflammatory diseases. Research into LILRA5 aims to uncover its specific ligands and signaling pathways, with potential implications for therapeutic targeting in immune-related disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA5
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LILRA5
Gene Accession No. 0
Gene Alias CD85; CD85 antigen-like family member F; CD85f; CD85f antigen; E030017K20; EG232801; Gm4878; ILT11; ILT-11; Immunoglobulin-like transcript 11; immunoglobulin-like transcript 11 protein; leucocyte Ig-like receptor A5; leukocyte Ig-like receptor 9; leukocyte immunoglobulin like receptor A5; leukocyte immunoglobulin-like receptor; leukocyte immunoglobulin-like receptor 9; Leukocyte immunoglobulin-like receptor subfamily A member 5; leukocyte immunoglobulin-like receptor subfamily A member 5 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 7; LILRA5; LILRB7; Lilrc1; LIR9; LIR-9
Gene Symbols LILRA5
Host Species Mouse
Immunogen Recombinant Rat LILRA5 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 691533
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.