Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LILRA5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LILRA5 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2241
![]() ![]() |
Proteintech Group Inc
188211AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody recognizes the isoform 1 and isoform 2 of LILRA5.
May play a role in triggering innate immune responses. Does not seem to play a role for any class I MHC antigen recognition.Specifications
LILRA5 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
A6NI73 | |
353514 | |
Peptide | |
Primary | |
-20°C | |
LILRA5 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD85, CD85F, ILT 11, ILT11, LILRA5, LILRB7, LIR 9, LIR9 | |
LILRA5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title