Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LILRA5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571561
Description
May play a role in triggering innate immune responses. Does not seem to play a role for any class I MHC antigen recognition.
Specifications
LILRA5 | |
Polyclonal | |
Unconjugated | |
LILRA5 | |
CD85f; ILT11; LILRB7; LIR9; CD85; CD85 antigen-like family member F; CD85F; CD85f antigen; ILT-11; ILT11CD85f; Immunoglobulin-like transcript 11; leukocyte Ig-like receptor 9; Leukocyte immunoglobulin-like receptor 9; leukocyte immunoglobulin-like receptor subfamily A member 5; leukocyte immunoglobulin-like receptor subfamily A member 5 soluble; leukocyte immunoglobulin-like receptor; subfamily A (with TM domain); member 5; leukocyte immunoglobulin-like receptor; subfamily B (with TM and ITIM domains); LILRB7; LIR-9; LIR9immunoglobulin-like transcript 11 protein; member 7 | |
Rabbit | |
Protein A | |
RUO | |
353514 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.63 mg/mL | |
PBS with 0.09% sodium azide | |
A6NI73 | |
LILRA5 | |
KLH conjugated synthetic peptide between 56-85 amino acids from the N-terminal region of human LILRA5 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction