Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA6 Monoclonal Antibody (1B5B11)
GREENER_CHOICE

Catalog No. PIMA538559
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538559 Supplier Invitrogen™ Supplier No. MA538559
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

LILRA6 may act as receptor for class I MHC antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA6
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1B5B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene LILRA6
Gene Accession No. Q6PI73
Gene Alias CD85b; ILT5; ILT8; ILT-8; immunoglobulin-like transcript 5; immunoglobulin-like transcript 8; leucocyte Ig-like receptor A6; leukocyte Ig-like receptor; leukocyte immunoglobulin like receptor A6; leukocyte immunoglobulin-like receptor subfamily A member 6; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 6; LILRA6; LILRB3; LILRB6
Gene Symbols LILRA6
Host Species Mouse
Immunogen Purified recombinant fragment of human LILRA6 (aa: 52-195) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.