Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRB3 Monoclonal Antibody (8A11F10A4)
GREENER_CHOICE

Catalog No. PIMA530648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530648 Supplier Invitrogen™ Supplier No. MA530648
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LILRB3.

This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRB3
Applications ELISA
Classification Monoclonal
Clone 8A11F10A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LILRB3
Gene Accession No. O75022
Gene Alias CD85 antigen-like family member A; CD85A; Cell-surface glycoprotein p91; Gp91; HL9; ILT5; ILT-5; immunoglobulin-like transcript 5; leucocyte Ig-like receptor B3; leukocyte immunoglobulin like receptor B3; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRA6; Lilrb3; LIR3; LIR-3; monocyte inhibitory receptor HL9; paired Ig-like receptor B; paired immunoglobulin-like receptor B; PIRB; PIR-B
Gene Symbols LILRB3
Host Species Mouse
Immunogen Recombinant Human LILRB3 Protein (Met1-Glu443).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11025
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.