Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LILRB3 Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc LILRB3312AP

Description
LILRB3 Polyclonal antibody specifically detects LILRB3 in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
LILRB3 | |
Polyclonal | |
Unconjugated | |
LILRB3 | |
CD85 antigen-like family member A; CD85A; Cell-surface glycoprotein p91; Gp91; HL9; ILT5; ILT-5; immunoglobulin-like transcript 5; leucocyte Ig-like receptor B3; leukocyte immunoglobulin like receptor B3; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRA6; Lilrb3; LIR3; LIR-3; monocyte inhibitory receptor HL9; paired Ig-like receptor B; paired immunoglobulin-like receptor B; PIRB; PIR-B | |
Synthetic peptide taken within amino acid region 580-631 on human LILRB3 protein. | |
200 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
O75022 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11025, 450147 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction