Learn More
Invitrogen™ LILRB4 Polyclonal Antibody

Description
LILRB4 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | LILRB4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LILRB4 |
| Gene Accession No. | Q8NHJ6 |
| Gene Alias | CD85 antigen-like family member K; CD85K; glycoprotein 49 B; glycoprotein 49b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; mast cell surface glycoprotein Gp49B; Monocyte inhibitory receptor HM18 |
| Gene Symbols | LILRB4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.