Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LILRB4 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30282
Description
Rabbit polyclonal antibody raised against partial recombinant human LILRB4.
- High specificity
- High reactivity
Specifications
LILRB4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant human LILRB4. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
CD85K/HM18/ILT3/LILRB5/LIR-5/LIR5 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
11006 | |
Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50 - 1:200) The optimal working dilution should be determined by the end user. | |
Q8NHJ6 | |
LILRB4 | |
Recombinant protein corresponding to amino acids 213-237 of human LILRB4. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction