Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552906 Supplier Invitrogen™ Supplier No. PA552906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKPTLWAEPA SVIARGKPVT LWCQGPLETE EYRLDKEGLP WARKRQNPLE PGAKAKFHIP STVYDSAGRY RCYYETPAGW SEPSDPLELV ATG Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.

This gene is a member of the leukocyte immunoglobulin-likereceptor (LIR) family, which is found in a gene cluster atchromosomal region 19q13.4. The encoded protein belongs to thesubfamily B class of LIR receptors which contain two or fourextracellular immunoglobulin domains, a transmembrane domain, andtwo to four cytoplasmic immunoreceptor tyrosine-based inhibitorymotifs (ITIMs). Several other LIR subfamily B receptors areexpressed on immune cells where they bind to MHC class I moleculeson antigen-presenting cells and inhibit stimulation of an immuneresponse. Multiple transcript variants encoding different isoformshave been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRB5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LILRB5
Gene Accession No. O75023
Gene Alias CD85 antigen-like family member C; CD85C; leucocyte Ig-like receptor B5; leukocyte immunoglobulin like receptor B5; leukocyte immunoglobulin-like receptor 8; Leukocyte immunoglobulin-like receptor subfamily B member 5; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 5; LILRB5; LIR8; LIR-8
Gene Symbols LILRB5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LILRB5. Recombinant protein control fragment (Product #RP-90059).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.