Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM Kinase 1/2, p Thr508 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200073034 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254645 0.1 mg
NB413822 0.025 mg
NBP25464505 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP254645 Supplier Novus Biologicals Supplier No. NBP254645
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIM Kinase 1/2 Polyclonal specifically detects LIM Kinase 1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen LIM Kinase 1/2 (p Thr508)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Alias EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1
Gene Symbols LIMK1
Host Species Rabbit
Immunogen A peptide sequence around phosphorylation site of threonine 508 (R-Y-T (p) -V-V) derived from Human LIMK1
Molecular Weight of Antigen 72 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3984
Test Specificity This antibody detects endogenous level of LIM Kinase 1/2 only when phosphorylated at threonine 508.
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.