Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIM Kinase 1 Antibody (OTI3H6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271141MFV610
Description
LIM Kinase 1 Monoclonal antibody specifically detects LIM Kinase 1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow CytometrySpecifications
LIM Kinase 1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Flow Cytometry | |
OTI3H6 | |
Flow Cytometry | |
EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
Full length human recombinant protein of human LIMK1(NP_002305) produced in HEK293T cell. | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
3984 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction