Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIM Kinase 1 Antibody (SR2175), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32190250UL
Description
LIM Kinase 1 Monoclonal antibody specifically detects LIM Kinase 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
LIM Kinase 1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
SR2175 | |
Western Blot 1:500-1:2000 | |
EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
A synthesized peptide derived from human LIM Kinase 1 (Uniprot #: P53667) | |
50 μg | |
Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
3984 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction