Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675207
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-207 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-207 Supplier Invitrogen™ Supplier No. PA141588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human muscle tissue, human skeletal muscle protein.

This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Lhx1
Gene Accession No. P48742
Gene Alias hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein
Gene Symbols Lhx1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 149-165, 175-182, and 395-412 of human Lim-1.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.