Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984195
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4195 Supplier Bioss Supplier No. BS18272R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an eye lens-specific protein found at the junctions of lens fiber cells, where it may contribute to cell junctional organization. It acts as a receptor for calmodulin, and may play an important role in both lens development and cataractogenesis. Mutations in this gene have been associated with cataract formation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen LIM2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LIM2
Gene Accession No. P55344
Gene Alias 19kDa; 4833403J20; CTRCT19; Lens fiber membrane intrinsic protein; lens intrinsic membrane protein 2; lens intrinsic membrane protein 2, 19kDa; Lim2; MP17; MP18; MP19; MP20; To3; total opacity 3
Gene Symbols LIM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LIM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3982
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.