Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMA1 Mouse anti-Human, Clone: OTI1A3, TrueMAB™, Origene

Mouse Monoclonal Antibody
Supplier: OriGene TA808537S

This item is not returnable.
View return policy
Description
LIMA1 Monoclonal antibody specifically detects LIMA1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
LIMA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UHB6 | |
Mouse | |
Affinity chromatography | |
RUO | |
51474 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1A3 | |
Unconjugated | |
LIMA1 | |
1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
Human recombinant protein fragment corresponding to amino acids 1-275 of human LIMA1 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction