Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIMCH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552068 Supplier Invitrogen™ Supplier No. PA552068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTEEVKLIVT CNMRAQESEP VEGGLRKVPD LHKDDLAQQR IQGSLAPHRE PPSFITLSNI TEADLETWER LKVSEKARDG DVQHICASEP SPEIKAETAI RDDFANRKAR ASKKASSPRQ KFVHFGPVTE LDQQKW Highest antigen sequence identity to the following orthologs: Mouse - 22%, Rat - 24%.

Actin stress fibers-associated protein that activates non-muscle myosin IIa. Activates the non-muscle myosin IIa complex by promoting the phosphorylation of its regulatory subunit MRLC/MYL9. Through the activation of non-muscle myosin IIa, positively regulates actin stress fibers assembly and stabilizes focal adhesions. It therefore negatively regulates cell spreading and cell migration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMCH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIMCH1
Gene Accession No. Q9UPQ0
Gene Alias 3732412D22Rik; Kiaa1102; LIM and calponin homology domains 1; LIM and calponin homology domains-containing protein 1; Limch1; LIMCH1A; LMO7B; mKIAA1102
Gene Symbols LIMCH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LIMCH1. Recombinant protein control fragment (Product #RP-102346).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.