Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171937
Description
LIMD1 Polyclonal specifically detects LIMD1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
LIMD1 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
LIM domain-containing protein 1, LIM domains containing 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg | |
NP_055055.1 | |
LIMD1 | |
The immunogen for this product maps to a region between residue 50 and 100 of human LIM Domains Containing 1 using the numbering given in entry NP_055055.1 (GeneID 8994). | |
0.1 mL | |
Epigenetics | |
8994 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction