Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIMD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109915 Supplier Invitrogen™ Supplier No. PA5109915
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDKYDDLGLE ASKFIEDLNM YEASKDGLFR VDKGAGNNPE FEETRRVFAT KMAKIHLQQQ QQQLLQEETL PRGSRGPVNG GGRLGPQARW EVVGSKLTVD GAAKPPLAAS TGAPGAVTTL AAGQPPYPPQ EQRSRPYLHG TRHGSQDCGS RESLATSEMS AFHQPGPCED PSCLTHGDYY DNLSLASPKW GDKPGVSPSI.

The Zyxin family of proteins contains five members: Ajuba, Limd1, LPP, TRIP6 and Zyxin. Limd1 (LIM domain-containing protein 1) is a ubiquitously expressed tumor suppressor containing three LIM zinc-binding domains. LIM domains consist of a cysteine-rich consensus sequence containing two distinct zinc-binding subdomains, which mediate protein-protein interactions. Limd1 interacts with the proteins SQSTM1, Rb, p62 and TRAF6. Limd1 was first identified when the deletion of its gene was noted in some cervical cancers. Limd1 blocks in vitro and in vivo tumor growth and is downregulated in lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LIMD1
Gene Accession No. Q9QXD8, Q9UGP4
Gene Alias AW822033; D9Ertd192e; LIM domain-containing protein 1; LIM domains containing 1; Limd1
Gene Symbols LIMD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human LIMD1 (NP_055055.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29806, 8994
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.