Learn More
LIMD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LIMD1 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5814
![]() ![]() |
Proteintech Group Inc
281061AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Proteintech's 28106-1-AP antibody was generated by 133 amino acids, it could recognize the full-length protein in applications.
The Zyxin family of proteins contains five members: Ajuba, Limd1, LPP, TRIP6 and Zyxin. Limd1 (LIM domain-containing protein 1) is a ubiquitously expressed tumor suppressor containing three LIM zinc-binding domains. LIM domains consist of a cysteine-rich consensus sequence containing two distinct zinc-binding subdomains, which mediate protein-protein interactions. Limd1 interacts with the proteins SQSTM1, Rb, p62 and TRAF6. Limd1 was first identified when the deletion of its gene was noted in some cervical cancers. Limd1 blocks in vitro and in vivo tumor growth and is downregulated in lung cancer.Specifications
LIMD1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UGP4 | |
8994 | |
LIMD1 Fusion Protein Ag27974 | |
Primary | |
-20°C | |
LIMD1 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
LIM domains containing 1, LIMD1 | |
LIMD1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.