Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIME Antibody (mLIME-05), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147230AF750
Description
LIME Monoclonal specifically detects LIME in Mouse samples. It is validated for Western Blot.Specifications
LIME | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
LIME1 | |
Recombinant protein corresponding to amino acids 86-202 of mouse LIME | |
0.1 mL | |
Primary | |
Mouse monoclonal antibody mLIME-05 reacts with the cytoplasmic domain of LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed mainly by T cells. | |
Store at 4°C in the dark. |
Western Blot | |
mLIME-05 | |
Western Blot | |
dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067 | |
Mouse | |
Protein A purified | |
Signal Transduction | |
54923.0 | |
Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction