Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIME Antibody (mLIME-05) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP147230

Catalog No. NBP147230


Only null left
Add to Cart

Description

Description

LIME Monoclonal specifically detects LIME in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

LIME
Monoclonal
1 mg/mL
Western Blot 1 to 5 μg/mL, Immunoprecipitation 1:50
LIME1
Recombinant protein corresponding to amino acids 86-202 of mouse LIME
Protein A purified
RUO
Primary
Mouse monoclonal antibody mLIME-05 reacts with the cytoplasmic domain of LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed mainly by T cells.
Store at 4C. Do not freeze.
IgG2a
Western Blot, Immunoprecipitation
mLIME-05
Unconjugated
dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067
Mouse
30 kDa
0.1 mg
Signal Transduction
54923
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only