Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIME Antibody (OTI5D3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271974IR
Description
LIME Monoclonal specifically detects LIME in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LIME | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LIME1 | |
Recombinant protein fragment corresponding to amino acids 30-295 of human LIME (NP_060276) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5D3 | |
Western Blot, Immunohistochemistry | |
dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
54923.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction