Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIME Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589384
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589384 Supplier Invitrogen™ Supplier No. PA589384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGLPVSWAPP ALWVLGCCAL LLSLWALCTA CRRPEDAVAP RKRARRQRAR LQGSATAAEA SLLRRTHLCS LSKSDTRLHE LHRGPRSSRA LRPASMDLLR PHWLEVSRDI TGPQAAPSAF PHQELPRALP AAAATAGCAG LEATYSNVGL AALPGVSLAA SPVVAEYARV QKRKGTHRSP QEPQQGKTEV TPAAQVDVLY SRVCKPKRRD PGPTTDPLDP KGQGAILALA GDLAYQTLPL RALDVDSGPL ENVYESIREL GDPAGRSSTC GAGTPPASSC PSLGRGWRPL PASLP; Positive Samples: K-562, Mouse lung; Cellular Location: Cell membrane, Single-pass type III membrane protein.

LIME (Lck-interacting molecule) is a 30 kDa double-palmitoylated protein with unusually basic cytoplasmic domain, expressed by T cells. After ligation of CD4 or CD8 T cell coreceptors, LIME is phosphorylated by Src-family kinases and associates with Lck and Fyn kinases and with their negative regulator Csk. Interestingly, Csk-mediated phosphorylation of C-terminal negative-regulatory tyrosine of LIME-associated Lck can result in increase of enzymatic activity compared with the total pool of Lck, thus, LIME serves as a positive regulator of TCR-dependent T cell signaling. However, under some circumstances, LIME may mediate inhibitory signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIME
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Lime1
Gene Accession No. Q9EQR5, Q9H400
Gene Alias 2810038K19Rik; dJ583P15.4; FLJ20406; Lck interacting transmembrane adaptor 1; lck-interacting membrane protein; Lck-interacting molecule; lck-interacting transmembrane adapter 1; Lime; lime 1; LIME1; LP8067; RP4-583P15.5
Gene Symbols Lime1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-295 of human LIME1 (NP_0602762).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54923, 72699
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.