Learn More
Eisco™ Limestone Specimen
SCI_ED
Catalog No. p-7076837 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24726 1 Ea. 2 to 3 cm
S26591 10/Pk. 2 to 3 cm
S24727 12/Pk. 2 to 3 cm
S26677 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S24726 Supplier Eisco™ Supplier No. ESNG0033
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Limestone Specimen is made up of mostly calcium carbonate and organic material.
  • Limestone formed from precipitation of calcium carbonate out of solution, in ocean or in shallow marine environment
  • For use in demonstrating tendency of calcium carbonate to effervesce in acid
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Massive, gray variety is often found deposited in marine environments
  • Limestone is important rock in industry and education
Specifications
Product Type Sedimentary Rock, Limestone
For Use With (Application) Demonstrating tendency of calcium carbonate to effervesce in acid
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm