Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Limitin/IFN-zeta Antibody (RM0123-7D19), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212426AF405
Description
Limitin/IFN-zeta Monoclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Limitin/IFN-zeta | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Ifnz | |
Mouse recombinant Limitin/IFN-zeta | |
0.1 mL | |
319146 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0123-7D19 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
6030405N23Rik, BGIF, Ifnz, interferon zeta, Lmtn | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction