Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Limitin/IFN-zeta Antibody (RM0123-7D19) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen Limitin/IFN-zeta
Clone RM0123-7D19
Concentration LYOPH
Applications Blocking Assay, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019211
SDP
View Documents
Novus Biologicals
NBP2124260.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP212426
SDP
View Documents
Novus Biologicals
NBP212426
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Limitin/IFN-zeta Monoclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.
Specifications

Specifications

Limitin/IFN-zeta
LYOPH
Monoclonal
Purified
RUO
BAA83749
319146
IgG2
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
RM0123-7D19
Blocking Assay, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rat
Mouse
6030405N23Rik, BGIF, Ifnz, interferon zeta, Lmtn
Ifnz
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.