Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMK1 Monoclonal Antibody (OTI 7C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503007S


Description
LIMK1 Monoclonal antibody specifically detects LIMK1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
LIMK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75688 | |
Mouse | |
Affinity chromatography | |
RUO | |
5495 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI 7C3 | |
Unconjugated | |
PPM1B | |
EC 2.7.11.1; kinase LIMK1; KIZ-1; LIM domain kinase 1; lim kinase 1; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK1; LIMK-1 | |
Full length human recombinant protein of human PPM1B produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction