Learn More
LIMK1 Monoclonal Antibody (OTI2A11), TrueMAB™, OriGene
SDP
Mouse Monoclonal Antibody
Supplier:  OriGene TA503122
Encompass_Preferred
 View more versions of this product
Catalog No. 50-167-4088
Only null left
Add to Cart
Description
Description
There are approximately 40 known eukaryotic LIM proteins, so named for the LIM domains they contain. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.
Specifications
Specifications
LIMK1
Monoclonal
Unconjugated
LIMK1
LIMK, LIMK-1
Mouse
Affinity Chromatography
RUO
3984
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2A11
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P53667
LIMK1
Full length human recombit protein of human LIMK1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.