Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LIMKi 3
GREENER_CHOICESDP

Catalog No. 474510 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-451-0 Supplier R&D Systems™ Supplier No. 4745/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent LIM kinase inhibitor; antitumor

LIMKi 3 is a potent LIM kinase inhibitor (IC50 values are 7 and 8 nM for LIMK1 and LIMK2 respectively). Inhibits cofilin phosphorylation in MDA-MB-231 breast cancer cells. Reduces MDA-MB-231 tumor cell invasion in a 3D matrigel invasion assay.

Specifications

Quantity 10 mg
Product Line Tocris
Product Type LIMKi 3
Purity >98%
Molecular Weight (g/mol) 431.29
Inhibitors LIMKi 3

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.