Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Limonin

Catalog No. AC465470010
Click to view available options
Quantity:
250 mg
1 g

CAS: 1180-71-8 | C26H30O8 | 470.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1180-71-8
Molecular Weight (g/mol) 470.52
MDL Number MFCD00075922

Specifications

Alpha Vector LIMONIN
Melting Point 298°C to 300°C
Color White
Loss on Drying ≤5 %
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C26 H30 O8
Quantity 1 g
Formula Weight 470.52
Physical Form Powder
Chemical Name or Material Limonin
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.