Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ LIMPII/SR-B2 Antibody Blocking Peptide

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 950 |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB400-129 |
Gene Alias | AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII |
Gene Symbol | SCARB2 |
Quantity | 0.1 mg |
Research Category | Cellular Markers, Cholesterol Metabolism, Lipid and Metabolism, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction