Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIMPII/SR-B2 Antibody (SR2211), Novus Biologicals™
SDP

Catalog No. p-200006333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259019 100 μg
NB259850 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259019 Supplier Novus Biologicals Supplier No. NBP322186100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LIMPII/SR-B2 Monoclonal antibody specifically detects LIMPII/SR-B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen LIMPII/SR-B2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone SR2211
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII
Host Species Rabbit
Immunogen A synthesized peptide derived from human LIMPII/SR-B2 (Uniprot #: Q14108)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Cholesterol Metabolism, Lipid and Metabolism, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 950
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.