Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMPII/SR-B2 Rabbit anti-Human, Mouse, Rat, Clone: 7D2P5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | LIMPII/SR-B2 |
---|---|
Clone | 7D2P5 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
LIMPII/SR-B2 Monoclonal antibody specifically detects LIMPII/SR-B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
LIMPII/SR-B2 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SR-B2/LIMPII/SR-B2 (Q14108). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7D2P5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cellular Markers, Cholesterol Metabolism, Lipid and Metabolism, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
950 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title