Learn More
Invitrogen™ LIMS1 Monoclonal Antibody (5G7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515876
Description
MA5-15876 targets PINCH in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human samples. The MA5-15876 immunogen is purified recombinant fragment of human PINCH expressed in E. Coli. . MA5-15876 detects PINCH which has a predicted molecular weight of approximately 37kDa.
The protein encoded by this gene is an adaptor protein which contains five LIM domains, or double zinc fingers. The protein is likely involved in integrin signaling through its LIM domain-mediated interaction with integrin-linked kinase, found in focal adhesion plaques. It is also thought to act as a bridge linking integrin-linked kinase to NCK adaptor protein 2, which is involved in growth factor receptor kinase signaling pathways. Its localization to the periphery of spreading cells also suggests that this protein may play a role in integrin-mediated cell adhesion or spreading. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
LIMS1 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P48059 | |
LIMS1 | |
Purified recombinant fragment of human PINCH expressed in E. Coli. | |
RUO | |
3987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
5G7 | |
Unconjugated | |
LIMS1 | |
2310016J22Rik; 4921524A02Rik; AI507642; AU021743; AW551584; C430041B13Rik; LIM and senescent cell antigen-like domains 1; LIM and senescent cell antigen-like-containing domain protein 1; LIM zinc finger domain containing 1; LIMS1; Lims1l; LIM-type zinc finger domains 1; Particularly interesting new Cys-His protein 1; PINCH; PINCH1; PINCH-1; renal carcinoma antigen NY-REN-48; RGD1560732; senescent cell antigen | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.