Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMS1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LIMS1 |
---|---|
Concentration | 0.63 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6116
![]() ![]() |
Proteintech Group Inc
553361AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Pinch, also designated particularly interesting new Cys-His protein or NY-REN-48, is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by a PINCH-1 related protein PINCH-2, which also forms a complex with ILK.Specifications
LIMS1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P48059 | |
3987 | |
Peptide | |
Primary | |
-20°C | |
LIMS1 |
0.63 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LIMS1, PINCH, PINCH 1, PINCH1 | |
LIMS1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title