Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMS2 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30469220UL
Description
LIMS2 Polyclonal specifically detects LIMS2 in Human, Rat samples. It is validated for Western Blot.Specifications
LIMS2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
55679 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
FLJ10044, ILK-binding protein, LIM and senescent cell antigen-like domains 2, LIM and senescent cell antigen-like-containing domain protein 2, LIM-like protein 2, Particularly interesting new Cys-His protein 2, PINCH2, PINCH-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-341 of human LIMS2 (NP_001154875.1). | |
20 μL | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction