Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIMS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571143 Supplier Invitrogen™ Supplier No. PA571143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 86%; Horse: 86%; Human: 100%; Mouse: 92%; Rat: 92%.

LIMS3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMS3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIMS3
Gene Accession No. P0CW19
Gene Alias LIM and senescent cell antigen-like domains 3; LIM and senescent cell antigen-like-containing domain protein 3; LIM and senescent cell antigen-like-containing domain protein 3-like; LIM and senescent cell antigen-like-containing domain protein 4; LIM zinc finger domain containing 3; LIMS3; LIMS3L; LIMS4; LIM-type zinc finger domains 3; Particularly interesting new Cys-His protein 3; pinch 2; PINCH3; PINCH-3
Gene Symbols LIMS3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human LIMS3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 96626
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.