Learn More
FUJIFILM Wako Limulus Color KY Single Test
Catalog No. 12333025
Encompass
Change view
Click to view available options
Quantity:
25 Test Vials
Catalog No. Quantity
12-333-025 25 Test Vials
1 options
Catalog No. 12-333-025 Supplier FUJIFILM Wako Supplier No. 29153601
Add to Cart
Add to Cart
Quantitative kinetic chromogenic assays (KCA) that are endotoxin-specific and unreactive to (1,3)-ß-D-glucan, come with pre-dispensed LAL reagent for single measurement, ideal for testing smaller number of samples or less experienced user
  • KCA quantitative range detection limit of 0.0002 to 5 EU/mL
  • Reagent reacts with endotoxin over wide range of concentrations
  • High sensitivity to reduce chance of interfering factors
  • Can be used with Toxinometer™
  • Reduces chance of contamination and no wasted reagent during testing
Specifications
Form Lyophilized
Fill Volume 200 μL
Includes LAL Reagent
Quantity 25 Test Vials