Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN-28B Antibody (SR1509), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321675100UL
Description
LIN-28B Monoclonal antibody specifically detects LIN-28B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
LIN-28B | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR1509 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:50 | |
CSDD2protein lin-28 homolog B, FLJ16517, lin-28 homolog B (C. elegans), Lin-28.2, lin-28B | |
A synthesized peptide derived from human LIN-28B (Uniprot #: Q6ZN17) | |
100 μg | |
Epigenetics, Neuroscience, Stem Cell Markers | |
389421 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction