Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
lin-54 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214854H
Description
lin-54 Polyclonal antibody specifically detects lin-54 in Human samples. It is validated for Western BlotSpecifications
lin-54 | |
Polyclonal | |
Western Blot | |
CXCDC1, JC8.6, MIP120, Protein LIN-54 | |
This antibody maps to a region between residue 1 and 50 of human Lin-54 homolog using the numbering given in entry NP_919258.2 (GeneID 132660). | |
0.1 mL | |
Cell Cycle and Replication | |
132660 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction