Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
lin-54 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214854PE
Description
lin-54 Polyclonal antibody specifically detects lin-54 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| lin-54 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| CXCDC1, JC8.6, MIP120, Protein LIN-54 | |
| This antibody maps to a region between residue 1 and 50 of human Lin-54 homolog using the numbering given in entry NP_919258.2 (GeneID 132660). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 132660 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction