Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN28 Mouse anti-Human, Clone: 55CT58.12.1, Abnova™

Catalog No. 89934485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-485 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-485 Supplier Abnova Corporation Supplier No. MAB12278
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human LIN28.

Specifications

Antigen LIN28
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 55CT58.12.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human LIN28.
Dilution Immunohistochemistry (1:50-1:200) Immunofluorescence (1:10-1:50) Western Blot (1:200-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias CSDD1/FLJ12457/LIN-28/LIN28A/ZCCHC1
Gene Symbols LIN28
Host Species Mouse
Immunogen Recombinant protein corresponding to human LIN28.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79727
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.