Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lin28A/B Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8443R
Description
Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.Specifications
| Lin28A/B | |
| Polyclonal | |
| Unconjugated | |
| LIN28A | |
| AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; lin28; lin-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; Lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein lin-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; Testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 500562, 79727, 83557 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q8K3Y3, Q9H9Z2 | |
| LIN28A | |
| KLH conjugated synthetic peptide derived from human Lin28A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction