Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN28A Monoclonal Antibody (A4-A6), Huabio

Catalog No. 502544481
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4481 Supplier HUABIO RESEARCH INC Supplier No. M13011
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIN28A Monoclonal antibody specifically detects LIN28A in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen LIN28A
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A4-A6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene LIN28A
Gene Accession No. Q8K3Y3, Q9H9Z2
Gene Alias AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; Lin28; LIN-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein LIN-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1
Host Species Mouse
Immunogen Recombinant protein
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79727, 83557
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.