Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN28B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557745 Supplier Invitrogen™ Supplier No. PA557745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKCHYCQSIM HMVANCPHKN VAQPPASSQG RQEAESQPCT STLPREVGGG HGCTSPPFPQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN28B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIN28B
Gene Accession No. Q6ZN17
Gene Alias 2810403D23Rik; CSDD2; D030047M17Rik; lin-28 homolog B; lin-28 homolog B (C. elegans); Lin-28.2; Lin28b; Lin-28B; Protein lin-28 homolog B
Gene Symbols LIN28B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LIN28B. Recombinant protein control fragment (Product #RP-95030).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.