Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIN28B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571921
Description
LIN28B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Specifications
LIN28B | |
Polyclonal | |
Unconjugated | |
LIN28B | |
2810403D23Rik; CSDD2; D030047M17Rik; lin-28 homolog B; lin-28 homolog B (C. elegans); Lin-28.2; Lin28b; Lin-28B; Protein lin-28 homolog B | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
380669, 389421, 689054 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q45KJ6, Q6ZN17 | |
LIN28B | |
KLH conjugated synthetic peptide between 95-128 amino acids of human LIN28B. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction