Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIN28B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587187
Description
Purity is > 95% by SDS-PAGE.
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Specifications
LIN28B | |
Polyclonal | |
Unconjugated | |
LIN28B | |
2810403D23Rik; CSDD2; D030047M17Rik; lin-28 homolog B; lin-28 homolog B (C. elegans); Lin-28.2; Lin28b; Lin-28B; Protein lin-28 homolog B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
380669, 389421 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q45KJ6, Q6ZN17 | |
LIN28B | |
Recombinant fusion protein of human LIN28B. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction