Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIN28B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588890
Description
Immunogen sequence: VANCPHKNVA QPPASSQGRQ EAESQPCTST LPREVGGGHG CTSPPFPQEA RAEISERSGR SPQEASSTKS SIAPEEQSKK GPSVQKRKKT; Positive Samples: Mouse liver; Cellular Location: Cytoplasm, Nucleus, nucleolus.
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Specifications
LIN28B | |
Polyclonal | |
Unconjugated | |
LIN28B | |
2810403D23Rik; CSDD2; D030047M17Rik; lin-28 homolog B; lin-28 homolog B (C. elegans); Lin-28.2; Lin28b; Lin-28B; Protein lin-28 homolog B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
380669, 389421 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.28 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q45KJ6, Q6ZN17 | |
LIN28B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 161-250 of human LIN28B (NP_0010043171). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction