Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107157 Supplier Thermo Scientific Supplier No. PA5107157
Add to Cart
Add to Cart

LIN37 Polyclonal Antibody

LIN37 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN37
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene LIN37
Gene Accession No. Q96GY3
Gene Alias antolefinin; F25965; LIN37; lin-37; lin-37 DREAM MuvB core complex component; lin-37 homolog; MSTP064; protein F25965; protein lin-37 homolog; ZK418.4
Gene Symbols LIN37
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LIN37 (aa 71-121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.